Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199351.1 | NP_001186280.1 | 6 | 645 | ||
NM_001199352.1 | NP_001186281.1 | 6 | 509 | |||
NM_012040.3 | NP_036170.1 | 6 | 585 | |||
GenBank mRNA | AB023027.1 | - | 5 | 455 | ||
AF181984.1 | - | 6 | 559 | |||
AK134665.1 | - | 6 | 516 | |||
AK158755.1 | - | 6 | 510 | |||
BC051996.1 | - | 6 | 492 | |||
BC055891.1 | - | 6 | 485 | 4208888 | ||
CX243772.1 | - | 6 | 646 |