Search
Search

Cpt1a| Gene Symbol: Cpt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013495.2 | NP_038523.2 | 5 | 560 | ||
| GenBank mRNA | AF017175.1 | - | 448 | |||
| AK050213.1 | - | 5 | 853 | |||
| AK077454.1 | - | 5 | 514 | |||
| AK128925.1 | - | 5 | 514 | |||
| AK133472.1 | - | 5 | 515 | |||
| AK136487.1 | - | 5 | 514 | |||
| AK147770.1 | - | 5 | 515 | |||
| AK149688.1 | - | 5 | 511 | |||
| BC038395.1 | - | 4 | 495 | 2649352 | ||
| BC046383.1 | - | 4 | 495 | 4160676 | ||
| BC054791.1 | - | 5 | 541 | 6832731 | ||
| D49472.1 | - | 4 | 487 | |||