Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-Ke6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013543.2 | NP_038571.2 | 3, 4 | 397 | ||
GenBank mRNA | BC086927.1 | - | 3, 4 | 397 | 6775958 | |
CX243612.1 | - | 3, 4 | 366 | |||
KJ650217.1 | - | 3, 4 | 389 |