Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GJA1 SIRNA Assays ›
Gene Symbol:
GJA1
Gene Name:
gap junction protein alpha 1
Gene Aliases:
AVSD3, CMDR, CX43, EKVP, GJAL, HLHS1, HSS, ODDD, PPKCA
Chromosome Location:
Chr.6: 121435577 - 121449744 on Build GRCh38
UniGene ID:
Hs.700699
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
7111
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
FAM | 20 nmol
100 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
FAM | 20 nmol
100 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
FAM | 20 nmol
100 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GJA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000165.4
NP_000156.1
2
2027
GenBank mRNA
AK309490.1
-
3
856
BC026329.1
-
2
1843
4794131
X52947.1
-
2
1912
Back To Top
More Information
Panther Classification:
Molecular Function -
gap junction
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
in utero embryonic development
neuron migration
negative regulation of endothelial cell proliferation
heart looping
regulation of heart rate
epithelial cell maturation
lens development in camera-type eye
chronic inflammatory response
positive regulation of glomerular filtration
endothelium development
atrial ventricular junction remodeling
transport
apoptotic process
muscle contraction
signal transduction
positive regulation of cytosolic calcium ion concentration
cell-cell signaling
heart development
adult heart development
response to pH
response to glucose
vascular transport
positive regulation of gene expression
negative regulation of gene expression
cell communication by chemical coupling
cell communication by electrical coupling
positive regulation of cell communication by chemical coupling
ATP transport
gap junction assembly
negative regulation of cell growth
regulation of bone mineralization
positive regulation of insulin secretion
ion transmembrane transport
response to fluid shear stress
embryonic digit morphogenesis
positive regulation of I-kappaB kinase/NF-kappaB signaling
skeletal muscle tissue regeneration
response to peptide hormone
positive regulation of osteoblast differentiation
positive regulation of protein catabolic process
positive regulation of striated muscle tissue development
positive regulation of vasoconstriction
positive regulation of vasodilation
regulation of bone remodeling
blood vessel morphogenesis
neuron projection morphogenesis
protein oligomerization
regulation of calcium ion transport
negative regulation of cardiac muscle cell proliferation
milk ejection
regulation of ventricular cardiac muscle cell membrane repolarization
regulation of atrial cardiac muscle cell membrane depolarization
regulation of ventricular cardiac muscle cell membrane depolarization
negative regulation of wound healing
cellular response to mechanical stimulus
atrial cardiac muscle cell action potential
cell communication by electrical coupling involved in cardiac conduction
negative regulation of DNA biosynthetic process
regulation of bicellular tight junction assembly
positive regulation of behavioral fear response
signal transducer activity
receptor binding
gap junction channel activity
protein binding
ion transmembrane transporter activity
SH3 domain binding
PDZ domain binding
beta-tubulin binding
connexin binding
gap junction channel activity involved in cardiac conduction electrical coupling
scaffold protein binding
gap junction channel activity involved in cell communication by electrical coupling
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0