Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SOD2 SIRNA Assays ›
          Gene Symbol:
          Sod2
          Gene Name:
          superoxide dismutase 2, mitochondrial
          Gene Aliases:
          MnSOD, Sod-2
          Chromosome Location:
          Chr.17: 13007839 - 13018119 on Build GRCm38
          UniGene ID:
          Mm.290876
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 71474

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Sod2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013671.3 NP_038699.2 4, 5 729
          GenBank mRNA AK002428.1 - 4, 5 589
          AK002534.1 - 4, 5 590
          AK012354.1 - 4, 5 587
          AK135435.1 - 4, 5 718
          AK152047.1 - 4, 5 587
          AK167521.1 - 4, 5 590
          AK168082.1 - 4, 5 590
          AK168591.1 - 4, 5 721
          AK177471.1 - 349
          AK179113.1 - 349
          AK180974.1 - 349
          BC010548.1 - 4, 5 565 3581909
          BC018173.1 - 3, 4 510
          CT010273.1 - 4, 5 522
          X04972.1 - 4, 5 574
          Z18857.1 - 4, 5 557

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001024465, NM_001024466, NM_000636, Y00985, M36693, and others.

          Panther Classification:

          Molecular Function -

          oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          response to superoxide
          age-dependent response to oxidative stress
          age-dependent response to reactive oxygen species
          release of cytochrome c from mitochondria
          liver development
          detection of oxygen
          vasodilation by acetylcholine involved in regulation of systemic arterial blood pressure
          regulation of transcription from RNA polymerase II promoter
          glutathione metabolic process
          superoxide metabolic process
          response to oxidative stress
          mitochondrion organization
          heart development
          sensory perception of sound
          locomotory behavior
          regulation of blood pressure
          negative regulation of cell proliferation
          intrinsic apoptotic signaling pathway in response to DNA damage
          intrinsic apoptotic signaling pathway in response to oxidative stress
          apoptotic mitochondrial changes
          post-embryonic development
          response to gamma radiation
          response to activity
          removal of superoxide radicals
          respiratory electron transport chain
          hemopoiesis
          response to nutrient levels
          oxygen homeostasis
          vasodilation
          response to drug
          response to hydrogen peroxide
          superoxide anion generation
          hydrogen peroxide metabolic process
          negative regulation of apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of nitric oxide biosynthetic process
          negative regulation of fat cell differentiation
          negative regulation of fibroblast proliferation
          neuron development
          response to axon injury
          erythrophore differentiation
          hydrogen peroxide biosynthetic process
          regulation of catalytic activity
          protein homooligomerization
          protein homotetramerization
          regulation of mitochondrial membrane potential
          iron ion homeostasis
          response to hyperoxia
          oxidation-reduction process
          negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          DNA binding
          superoxide dismutase activity
          protein binding
          oxidoreductase activity
          oxygen binding
          manganese ion binding
          identical protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0