Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013753.2 | NP_038781.2 | 3 | 479 | ||
GenBank mRNA | AK088577.1 | - | 3 | 315 | ||
AK146489.1 | - | 3 | 423 | |||
BC052850.1 | - | 325 | ||||
BC080827.1 | - | 2 | 270 | |||
BC099531.1 | - | 3 | 294 | |||
BC137695.1 | - | 2 | 243 | 8860714 | ||
BC144846.1 | - | 2 | 249 | 9053394 | ||
BC144847.1 | - | 2 | 249 | 9053395 | ||
X99384.1 | - | 2 | 264 |