Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ip6k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013785.2 | NP_038813.2 | 4, 5 | 908 | ||
GenBank mRNA | AF177144.1 | - | 3, 4 | 608 | ||
AK037925.1 | - | 3, 4 | 718 | |||
AK088872.1 | - | 4, 5 | 925 | |||
AK171310.1 | - | 4, 5 | 913 | |||
AK172921.1 | - | 4, 5 | 967 | |||
BC053030.1 | - | 4, 5 | 913 | |||
BC059006.1 | - | 4, 5 | 908 | 6827203 |