Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164768.1 | NP_001158240.1 | 6 | 1755 | ||
NM_001164770.1 | NP_001158242.1 | 8 | 1722 | |||
NM_013890.4 | NP_038918.3 | 8 | 1814 | |||
GenBank mRNA | AF140683.1 | - | 6 | 1759 | ||
AK033129.1 | - | 8 | 1768 | |||
AK078815.1 | - | 8 | 1722 | |||
AK088898.1 | - | 5 | 1324 | |||
X54352.1 | - | 7 | 1743 |