Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130412.1 | NP_001123884.1 | 5 | 681 | ||
NM_015763.4 | NP_056578.2 | 4 | 537 | |||
NM_172950.3 | NP_766538.2 | 4 | 537 | |||
GenBank mRNA | AF180471.1 | - | 4 | 516 | ||
AF412811.1 | - | 3 | 475 | |||
AK014526.1 | - | 3 | 498 | |||
AK019539.1 | - | 4 | 675 | |||
AK030921.1 | - | 4 | 540 | |||
AK141489.1 | - | 4 | 502 | |||
AK154837.1 | - | 5 | 678 | |||
BC042462.1 | - | 3 | 491 | 4211202 |