Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015830.1 | NP_056645.1 | 1 | 656 | ||
GenBank mRNA | AK079422.1 | - | 2 | 889 | ||
AK143711.1 | - | 3 | 1295 | |||
AK170543.1 | - | 2 | 890 | |||
AK212570.1 | - | 1 | 97 | |||
BC058094.1 | - | 2 | 914 | 6824892 |