Search
Search

Gpc3| Gene Symbol: Gpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016697.3 | NP_057906.2 | 1 | 286 | ||
| GenBank mRNA | AF185614.1 | - | 1 | 226 | ||
| AK077942.1 | - | 1 | 264 | |||
| AK146702.1 | - | 1 | 263 | |||
| AK159394.1 | - | 1 | 255 | |||
| AK159435.1 | - | 1 | 255 | |||
| AK159764.1 | - | 1 | 263 | |||
| AK168402.1 | - | 1 | 264 | |||
| BC036126.1 | - | 1 | 233 | 4973409 | ||
| CJ166243.1 | - | 1 | 287 | |||