Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276710.1 | NP_001263639.1 | 2 | 302 | ||
NM_016702.3 | NP_057911.2 | 2 | 302 | |||
GenBank mRNA | AF027730.2 | - | 2 | 297 | ||
AK149432.1 | - | 2 | 225 | |||
BB868742.1 | - | 2 | 303 | |||
BC025799.1 | - | 2 | 266 | 5099627 |