Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016786.3 | NP_058066.2 | 2, 3 | 388 | ||
GenBank mRNA | AB011081.1 | - | 2, 3 | 326 | ||
AK018366.1 | - | 2, 3 | 362 | |||
AK035371.1 | - | 2, 3 | 285 | |||
AK036068.1 | - | 2, 3 | 329 | |||
AK048194.1 | - | 2, 3 | 379 | |||
AK139564.1 | - | 2, 3 | 405 | |||
AK160728.1 | - | 2, 3 | 348 | |||
AK162142.1 | - | 2, 3 | 329 | |||
AK166647.1 | - | 2, 3 | 297 | |||
BC002013.1 | - | 2, 3 | 236 | 3496393 | ||
BC085311.1 | - | 2, 3 | 314 | 30148009 | ||
BY247382.1 | - | 2, 3 | 389 |