Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016750.3 | NP_058030.1 | 5 | 629 | ||
GenBank mRNA | AK028176.1 | - | 5 | 526 | ||
AK088704.1 | - | 5 | 576 | |||
AK143572.1 | - | 5 | 528 | |||
AK146181.1 | - | 5 | 561 | |||
AK150454.1 | - | 5 | 528 | |||
AK151052.1 | - | 5 | 528 | |||
AK151462.1 | - | 5 | 457 | |||
AK151509.1 | - | 5 | 528 | |||
AK151868.1 | - | 5 | 528 | |||
AK152398.1 | - | 5 | 526 | |||
AK152848.1 | - | 5 | 527 | |||
AK168178.1 | - | 5 | 528 | |||
AK169003.1 | - | 5 | 414 | |||
BC079903.1 | - | 5 | 551 | 30622912 | ||
BC147478.1 | - | 5 | 466 | 9056200 | ||
BC158035.1 | - | 5 | 466 | 9007338 | ||
BC171999.1 | - | 5 | 466 | 9054285 | ||
U70494.1 | - | 5 | 517 |
Gene Symbol: Gm8203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK210116.1 | - | 1 | 63 |