Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ybx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016875.2 | NP_058571.2 | 3 | 435 | ||
GenBank mRNA | AF073954.1 | - | 3 | 362 | ||
AF073955.1 | - | 3 | 229 | |||
AK131786.1 | - | 3 | 413 | |||
BC100454.1 | - | 3 | 435 | |||
BC141112.1 | - | 3 | 538 | 9055724 | ||
DV661016.1 | - | 2 | 37 |