Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017376.3 | NP_059072.1 | 2 | 546 | ||
NM_153484.3 | NP_705617.2 | 2 | 453 | |||
GenBank mRNA | AF194420.1 | - | 2 | 480 | ||
AK042146.1 | - | 2 | 707 | |||
AK045575.1 | - | 2 | 402 | |||
AK053229.1 | - | 2 | 391 | |||
AK075601.1 | - | 1 | 104 | |||
AK141606.1 | - | 2 | 374 | |||
AK145125.1 | - | 2 | 515 | |||
AK147965.1 | - | 2 | 479 | |||
AK154472.1 | - | 2 | 491 | |||
AK177827.1 | - | 2 | 257 | |||
AK185529.1 | - | 2 | 257 | |||
AK185648.1 | - | 1 | 257 | |||
AK196541.1 | - | 1 | 77 | |||
AK200544.1 | - | 1 | 78 | |||
AK214482.1 | - | 2 | 257 | |||
AY540631.1 | - | 2 | 532 | |||
AY540632.1 | - | 2 | 347 | |||
BC017689.1 | - | 2 | 309 | 4242534 | ||
BC036982.1 | - | 2 | 510 | 4976241 |