Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320350.1 | NP_001307279.1 | 6 | 1019 | ||
NM_001320351.1 | NP_001307280.1 | 6 | 1019 | |||
NM_003307.3 | NP_003298.1 | 6 | 1019 | |||
NR_038257.1 | 6 | 1019 | ||||
XM_005261171.3 | XP_005261228.1 | 7 | 1180 | |||
XM_011529736.2 | XP_011528038.1 | 6 | 1423 | |||
XM_017028456.1 | XP_016883945.1 | 7 | 1183 | |||
XM_017028457.1 | XP_016883946.1 | 6 | 1424 | |||
GenBank mRNA | AB001535.1 | - | 6 | 1364 | ||
AB166745.1 | - | 7 | 1132 | |||
AB166746.1 | - | 2 | 371 | |||
AJ417076.1 | - | 6 | 932 | |||
AJ878416.1 | - | 6 | 982 | |||
AY603182.1 | - | 6 | 919 | |||
BC112342.1 | - | 6 | 932 | 40069893 |