Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018857.1 | NP_061345.1 | 4 | 335 | ||
GenBank mRNA | AK144391.1 | - | 5 | 267 | ||
AK146799.1 | - | 4 | 385 | |||
BC023753.1 | - | 4 | 339 | 5354484 | ||
D86370.1 | - | 4 | 335 |