Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018861.3 | NP_061349.3 | 4 | 907 | ||
GenBank mRNA | AF246129.1 | - | 4 | 666 | ||
AK044280.1 | - | 4 | 878 | |||
AK136900.1 | - | 4 | 907 | |||
AK139248.1 | - | 4 | 906 | |||
AK140866.1 | - | 4 | 908 | |||
AK148075.1 | - | 4 | 908 | |||
AK202085.1 | - | 2 | 49 | |||
AK213050.1 | - | 2 | 49 | |||
BC043483.1 | - | 4 | 865 | 5003744 | ||
BC052733.1 | - | 4 | 781 | 6816689 | ||
U75215.1 | - | 4 | 777 |