Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bri3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163709.1 | NP_001157181.1 | 3 | 594 | ||
NM_018772.4 | NP_061242.3 | 3 | 710 | |||
GenBank mRNA | AF106967.1 | - | 3 | 629 | ||
AK080467.1 | - | 3 | 700 | |||
BC005677.1 | - | 3 | 688 | |||
BC021149.1 | - | 3 | 656 | |||
BC095955.1 | - | 3 | 665 | 5328625 | ||
X61454.1 | - | 2 | 358 |