Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018858.2 | NP_061346.2 | 2, 3 | 260 | ||
GenBank mRNA | AB046417.1 | - | 2, 3 | 262 | ||
AF300422.1 | - | 2, 3 | 239 | |||
AK008037.1 | - | 276 | ||||
AK088212.1 | - | 2, 3 | 265 | |||
AK143897.1 | - | 2, 3 | 292 | |||
AK149815.1 | - | 2, 3 | 276 | |||
AK159842.1 | - | 2, 3 | 265 | |||
AK182328.1 | - | 2, 3 | 150 | |||
BC008169.1 | - | 2, 3 | 294 | 3257403 | ||
BC083063.1 | - | 2, 3 | 260 | 5362125 | ||
BC089332.1 | - | 2, 3 | 243 | 30359654 | ||
U43206.1 | - | 2, 3 | 258 |