Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018888.4 | NP_061376.2 | 5, 6 | 663 | ||
GenBank mRNA | AF253516.1 | - | 5, 6 | 410 | ||
AK010376.1 | - | 5, 6 | 404 | |||
AK011152.1 | - | 5, 6 | 417 | |||
AK011953.1 | - | 5, 6 | 385 | |||
AK077323.1 | - | 5, 6 | 401 | |||
AK139488.1 | - | 5, 6 | 663 | |||
AK169413.1 | - | 5, 6 | 400 | |||
BC050884.1 | - | 5, 6 | 616 | 6808756 | ||
BC057570.1 | - | 5, 6 | 616 | 6808756 |