Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019443.2 | NP_062316.1 | 1, 2 | 258 | ||
GenBank mRNA | AK007840.1 | - | 1, 2 | 165 | ||
AK131584.1 | - | 457 | ||||
BC024364.1 | - | 1, 2 | 167 | 5031206 | ||
CF383819.1 | - | 42 | ||||
CT010398.1 | - | 1, 2 | 110 | |||
Y07708.1 | - | 1, 2 | 277 |