Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204165.1 | NP_001191094.1 | 2, 3 | 279 | ||
NM_001204167.1 | NP_001191096.1 | 2, 3 | 279 | |||
NM_019688.2 | NP_062662.1 | 2, 3 | 279 | |||
GenBank mRNA | AB021132.1 | - | 2, 3 | 254 | ||
AF115480.1 | - | 2, 3 | 207 | |||
AK017479.1 | - | 2, 3 | 236 | |||
AK161934.1 | - | 2, 3 | 247 | |||
AK220281.1 | - | 2, 3 | 279 | |||
BC139822.1 | - | 2, 3 | 211 | 40141811 |