Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019919.3 | NP_064303.2 | 4 | 1323 | ||
GenBank mRNA | AF022889.1 | - | 4 | 1288 | ||
CV559283.1 | - | 3 | 458 |