Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019916.2 | NP_064300.2 | 1 | 526 | ||
GenBank mRNA | AJ223801.1 | - | 1 | 397 | ||
AK141870.1 | - | 1 | 527 | |||
BC141308.1 | - | 1 | 456 | 9055920 | ||
BC145631.1 | - | 1 | 456 | 9054199 |