Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039509.1 | NP_001034598.1 | 4, 5 | 510 | ||
NM_019999.2 | NP_064383.1 | 3, 4 | 419 | |||
GenBank mRNA | AB041609.1 | - | 3, 4 | 420 | ||
AK041238.1 | - | 3, 4 | 353 | |||
AK173125.1 | - | 4, 5 | 484 | |||
BC058945.1 | - | 5, 6 | 606 | 6812127 |