Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ick | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163780.1 | NP_001157252.1 | 12 | 1998 | ||
NM_019987.2 | NP_064371.2 | 12 | 1926 | |||
GenBank mRNA | AF225918.1 | - | 12 | 1766 | ||
AK052178.1 | - | 5 | 895 | |||
AK087484.1 | - | 12 | 1850 | |||
AK133537.1 | - | 12 | 1865 | |||
AK154185.1 | - | 12 | 1999 | |||
AK173068.1 | - | 7 | 1155 | |||
BC028863.1 | - | 11 | 1683 | 4224269 |