Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MTOR SIRNA Assays ›
          Gene Symbol:
          Mtor
          Gene Name:
          mechanistic target of rapamycin (serine/threonine kinase)
          Gene Aliases:
          2610315D21Rik, AI327068, FRAP, FRAP2, Frap1, RAFT1, RAPT1, flat
          Chromosome Location:
          Chr.4: 148448582 - 148557685 on Build GRCm38
          UniGene ID:
          Mm.21158
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 74973

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mtor
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_020009.2 NP_064393.2 6 887
          GenBank mRNA AF152838.1 - 6 862
          AK143381.1 - 3 298
          AK172584.1 - 6 878
          BC112904.1 - 6 845 6465126

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          regulation of glycogen biosynthetic process
          regulation of carbohydrate metabolic process
          protein phosphorylation
          response to stress
          germ cell development
          negative regulation of autophagy
          positive regulation of lamellipodium assembly
          positive regulation of gene expression
          positive regulation of myotube differentiation
          cell growth
          negative regulation of macroautophagy
          phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          cell projection organization
          positive regulation of actin filament polymerization
          ruffle organization
          regulation of myelination
          cellular response to nutrient levels
          TOR signaling
          regulation of fatty acid beta-oxidation
          regulation of response to food
          response to insulin
          regulation of actin cytoskeleton organization
          regulation of GTPase activity
          response to amino acid
          regulation of carbohydrate utilization
          regulation of osteoclast differentiation
          positive regulation of translation
          negative regulation of cell size
          regulation of protein kinase activity
          positive regulation of transcription from RNA polymerase III promoter
          protein autophosphorylation
          positive regulation of lipid biosynthetic process
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of stress fiber assembly
          negative regulation of NFAT protein import into nucleus
          positive regulation of protein kinase B signaling
          cellular response to hypoxia
          nucleotide binding
          RNA polymerase III type 1 promoter DNA binding
          RNA polymerase III type 2 promoter DNA binding
          RNA polymerase III type 3 promoter DNA binding
          TFIIIC-class transcription factor binding
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          drug binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          phosphotransferase activity, alcohol group as acceptor
          protein domain specific binding
          ribosome binding
          protein dimerization activity
          phosphoprotein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0