Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMBT1 SIRNA Assays ›
Gene Symbol:
DMBT1
Gene Name:
deleted in malignant brain tumors 1
Gene Aliases:
GP340, SAG, muclin
Chromosome Location:
Chr.10: 122560665 - 122643736 on Build GRCh38
UniGene ID:
Hs.279611
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
7499
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMBT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320644.1
NP_001307573.1
2, 3
191
NM_004406.2
NP_004397.2
2, 3
191
NM_007329.2
NP_015568.2
2, 3
191
NM_017579.2
NP_060049.2
2, 3
191
XM_006717660.3
XP_006717723.1
2, 3
179
XM_006717665.3
XP_006717728.1
2, 3
179
XM_011539388.2
XP_011537690.1
2, 3
179
XM_011539389.2
XP_011537691.1
2, 3
179
XM_011539390.2
XP_011537692.1
2, 3
179
XM_011539391.2
XP_011537693.1
2, 3
179
XM_011539392.2
XP_011537694.1
2, 3
179
XM_011539393.2
XP_011537695.1
2, 3
179
XM_011539394.2
XP_011537696.1
2, 3
179
XM_011539395.2
XP_011537697.1
2, 3
179
XM_011539396.2
XP_011537698.1
2, 3
179
XM_011539398.2
XP_011537700.1
2, 3
179
XM_011539399.2
XP_011537701.1
2, 3
179
XM_011539400.2
XP_011537702.1
2, 3
179
XM_011539401.2
XP_011537703.1
2, 3
179
XM_011539402.2
XP_011537704.1
2, 3
179
XM_011539403.2
XP_011537705.1
2, 3
179
XM_011539405.2
XP_011537707.1
2, 3
180
XM_011539407.2
XP_011537709.1
2, 3
179
XM_011539408.2
XP_011537710.1
2, 3
179
XM_011539409.2
XP_011537711.1
2, 3
179
XM_011539410.2
XP_011537712.1
2, 3
179
XM_011539411.2
XP_011537713.1
2, 3
179
XM_011539413.2
XP_011537715.1
2, 3
178
XM_011539414.2
XP_011537716.1
2, 3
176
XM_011539415.2
XP_011537717.1
2, 3
181
GenBank mRNA
AF159456.1
-
2, 3
191
AJ000342.1
-
2, 3
191
AJ243212.1
-
2, 3
191
AJ243224.1
-
2, 3
191
AJ297935.1
-
2, 3
172
AK304128.1
-
2, 3
101
AK304149.1
-
2, 3
171
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
pattern recognition receptor signaling pathway
receptor-mediated endocytosis
multicellular organism development
protein transport
epithelial cell differentiation
induction of bacterial agglutination
cellular protein metabolic process
innate immune response
defense response to virus
scavenger receptor activity
protein binding
signaling pattern recognition receptor activity
zymogen binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-zbtnl:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0