Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020332.4 | NP_065065.3 | 2 | 464 | ||
GenBank mRNA | AF001532.1 | - | 2 | 167 | ||
AF274752.1 | - | 2 | 167 | |||
AK083135.1 | - | 2 | 465 | |||
AK148070.1 | - | 2 | 237 | |||
BC042644.1 | - | 2 | 389 | |||
BC054379.1 | - | 2 | 371 | 6390764 | ||
DQ832285.1 | - | 2 | 464 |