Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020046.3 | NP_064430.1 | 9 | 1793 | ||
GenBank mRNA | AF029667.2 | - | 9 | 1498 | ||
AK013102.1 | - | 7 | 1440 | |||
AK030798.1 | - | 1377 | ||||
BC019542.1 | - | 9 | 1503 | 4039461 | ||
BC027829.1 | - | 9 | 1754 | 5253723 | ||
BC045206.1 | - | 9 | 1743 | 5253764 |