Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020332.4 | NP_065065.3 | 12 | 1948 | ||
GenBank mRNA | AF001533.2 | - | 10 | 1294 | ||
AF274752.1 | - | 12 | 1651 | |||
AK083135.1 | - | 12 | 1950 | |||
AK086014.1 | - | 5 | 819 | |||
AK148070.1 | - | 12 | 1721 | |||
BC042644.1 | - | 12 | 1871 | |||
BC054379.1 | - | 12 | 1855 | 6390764 | ||
DQ832285.1 | - | 12 | 1948 |