siRNA Details
Gene Symbol: Smarce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020618.4 | NP_065643.1 | 5, 6 | 893 | ||
GenBank mRNA | AK018493.1 | - | 5, 6 | 321 | ||
AK076177.1 | - | 5, 6 | 325 | |||
BC034509.1 | - | 5, 6 | 308 | |||
BC047141.1 | - | 5, 6 | 314 | 6397290 | ||
BC061498.1 | - | 5, 6 | 321 | 5251562 | ||
BC065043.1 | - | 5, 6 | 893 | 5685483 | ||
EU327022.1 | - | 4, 5 | 218 | |||
EU327023.1 | - | 3, 4 | 174 | |||
EU327024.1 | - | 4, 5 | 271 | |||
EU327025.1 | - | 3, 4 | 166 | |||
EU327026.1 | - | 2, 3 | 122 |

More Information
Other Species Targeted

Panther Classification:
