Search
Search

Htr6| Gene Symbol: Htr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021358.2 | NP_067333.1 | 3 | 940 | ||
| GenBank mRNA | AF134158.1 | - | 3 | 940 | ||
| BC116652.1 | - | 3 | 940 | 40104888 | ||
| BC116653.1 | - | 3 | 940 | 40104889 | ||
Set Membership: |
|