Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021353.3 | NP_067328.1 | 21 | 2513 | ||
GenBank mRNA | AF136751.1 | - | 21 | 2431 | ||
AK018617.1 | - | 1 | 939 |