Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017424.2 | NP_001017424.1 | 3 | 758 | ||
NM_001017425.2 | NP_001017425.2 | 3 | 559 | |||
NM_014217.3 | NP_055032.1 | 3 | 435 | |||
XM_011509522.2 | XP_011507824.1 | 3 | 1961 | |||
XM_011509523.1 | XP_011507825.1 | 3 | 363 | |||
XM_011509524.2 | XP_011507826.1 | 3 | 410 | |||
XM_017001248.1 | XP_016856737.1 | 4 | 1146 | |||
XM_017001249.1 | XP_016856738.1 | 4 | 1097 | |||
GenBank mRNA | AF004711.1 | - | 3 | 543 | ||
AF129399.1 | - | 2 | 345 | |||
AF171068.1 | - | 3 | 359 | |||
AK291483.1 | - | 3 | 539 | |||
AK315249.1 | - | 4 | 525 | |||
AU280191.1 | - | 3 | 501 | |||
AY552980.2 | - | 3 | 445 | |||
AY552981.1 | - | 3 | 450 | |||
BC069462.1 | - | 3 | 359 | 7262344 | ||
BC101693.1 | - | 3 | 404 | 8069199 | ||
BC101695.1 | - | 3 | 404 | 8069201 | ||
BC143586.1 | - | 3 | 405 | 9052097 | ||
CD625301.1 | - | 3 | 374 | |||
DR001358.1 | - | 3 | 570 | |||
EF165334.1 | - | 3 | 492 | |||
EF165335.1 | - | 3 | 457 | |||
KF182338.1 | - | 3 | 394 | |||
KF182339.1 | - | 3 | 368 |