Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159631.1 | NP_001153103.1 | 3 | 301 | ||
NM_021606.3 | NP_067619.1 | 3 | 294 | |||
GenBank mRNA | AF218847.1 | - | 3 | 296 | ||
AK004925.1 | - | 3 | 271 | |||
AK011519.1 | - | 3 | 280 | |||
AK018168.1 | - | 3 | 284 | |||
AK029266.1 | - | 3 | 283 | |||
AK075717.1 | - | 3 | 268 | |||
AK075836.1 | - | 3 | 280 | |||
AK086700.1 | - | 299 | ||||
AK089919.1 | - | 3 | 255 | |||
AK170757.1 | - | 3 | 301 | |||
BC019524.1 | - | 3 | 363 | 4209097 | ||
BY205493.1 | - | 3 | 301 |