Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iigp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146275.1 | NP_001139747.1 | 3 | 447 | ||
NM_021792.4 | NP_068564.4 | 2 | 264 | |||
GenBank mRNA | AF194871.1 | - | 1 | 159 | ||
AJ007971.1 | - | 2 | 246 | |||
AK089702.1 | - | 2 | 275 | |||
AK153060.1 | - | 2 | 266 | |||
BC004649.1 | - | 2 | 241 | 3483102 | ||
CT010300.1 | - | 1 | 142 | |||
DV061261.1 | - | 2 | 297 |