Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001248011.1 | NP_001234940.1 | 2 | 575 | ||
NM_024047.4 | NP_076952.1 | 2 | 575 | |||
NM_198038.2 | NP_932155.1 | 2 | 217 | |||
XM_011532031.2 | XP_011530333.1 | 2 | 578 | |||
XM_011532032.2 | XP_011530334.1 | 2 | 2754 | |||
GenBank mRNA | AF265341.1 | - | 2 | 216 | ||
AF273028.1 | - | 2 | 539 | |||
AK027295.1 | - | 2 | 565 | |||
AK074845.1 | - | 2 | 217 | |||
AK075408.1 | - | 2 | 495 | |||
AY026252.1 | - | 2 | 570 | |||
AY359123.1 | - | 2 | 269 | |||
BC000542.2 | - | 2 | 335 | 3163944 | ||
BI552154.1 | - | 215 | ||||
BM544951.1 | - | 2 | 587 |