Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022435.2 | NP_071880.1 | 2 | 1868 | ||
GenBank mRNA | AF279479.1 | - | 2 | 1625 | ||
AK036234.1 | - | 2 | 1868 |