Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lima1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113545.1 | NP_001107017.1 | 11 | 2565 | ||
NM_023063.4 | NP_075550.2 | 8 | 2437 | |||
GenBank mRNA | AF307844.1 | - | 8 | 2360 | ||
AK028186.1 | - | 8 | 2378 | |||
AK031698.1 | - | 8 | 2381 | |||
AK049350.1 | - | 11 | 2515 | |||
AK085065.1 | - | 8 | 2431 | |||
AK132314.1 | - | 8 | 2383 | |||
AK167220.1 | - | 8 | 1997 | |||
AK206767.1 | - | 1 | 22 | |||
BC031490.1 | - | 11 | 2491 | 3498138 |