Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023142.2 | NP_075631.2 | 10 | 1214 | ||
GenBank mRNA | AF162768.1 | - | 9 | 1139 | ||
AK002274.1 | - | 10 | 1215 | |||
AK005378.1 | - | 10 | 1215 | |||
AK147731.1 | - | 10 | 1182 | |||
AK148145.1 | - | 10 | 1208 | |||
AK151911.1 | - | 10 | 1214 | |||
AK153510.1 | - | 10 | 1214 | |||
AK157098.1 | - | 10 | 1205 | |||
AK159371.1 | - | 10 | 1213 | |||
AK159477.1 | - | 10 | 1104 | |||
AK167978.1 | - | 10 | 1144 | |||
AK168774.1 | - | 10 | 1212 | |||
AK170759.1 | - | 10 | 1204 | |||
AK171364.1 | - | 10 | 1176 | |||
AK172124.1 | - | 10 | 1182 | |||
AK192837.1 | - | 1 | 36 | |||
AK196003.1 | - | 1 | 36 | |||
AK206912.1 | - | 1 | 36 | |||
AK207221.1 | - | 1 | 36 | |||
BC003441.1 | - | 10 | 1146 | 2650641 | ||
BC010275.1 | - | 10 | 1156 | 3589768 | ||
BC020404.1 | - | 7 | 742 | |||
BC092051.1 | - | 10 | 1170 | 3668682 |