Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023712.3 | NP_076201.2 | 4 | 1357 | ||
GenBank mRNA | AF212372.1 | - | 4 | 848 | ||
AK033907.1 | - | 3 | 1827 | |||
AK154353.1 | - | 4 | 847 | |||
AK166931.1 | - | 4 | 849 | |||
AK193120.1 | - | 1 | 32 | |||
BC002297.1 | - | 4 | 376 | |||
BC026854.1 | - | 4 | 818 | 4242049 | ||
BC085491.1 | - | 4 | 801 | 6829415 |