Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023729.3 | NP_076218.3 | 3, 4 | 360 | ||
GenBank mRNA | AF459789.1 | - | 3, 4 | 354 | ||
AK016595.1 | - | 3, 4 | 355 | |||
BC058859.1 | - | 3, 4 | 369 | 30011651 | ||
BG088136.2 | - | 3, 4 | 360 |