Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289524.1 | NP_001276453.1 | 2 | 247 | ||
NM_024194.6 | NP_077156.2 | 2 | 247 | |||
GenBank mRNA | AK011726.1 | - | 2 | 228 | ||
AK017814.1 | - | 2 | 216 | |||
AK034978.1 | - | 227 | ||||
AK148748.1 | - | 2 | 206 | |||
AK163288.1 | - | 2 | 215 | |||
AK170052.1 | - | 2 | 227 | |||
CJ072347.1 | - | 2 | 250 |