Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025299.3 | NP_079575.1 | 4 | 551 | ||
GenBank mRNA | AK004039.1 | - | 4 | 508 | ||
AK007331.1 | - | 4 | 513 | |||
AK148287.1 | - | 4 | 534 | |||
AK166497.1 | - | 4 | 527 | |||
BC031634.1 | - | 4 | 492 | 3662554 |