Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000049.2 | NP_000040.1 | 1 | 299 | ||
NM_001128085.1 | NP_001121557.1 | 2 | 232 | |||
XM_017024661.1 | XP_016880150.1 | 3 | 448 | |||
GenBank mRNA | AK312901.1 | - | 2 | 290 | ||
BC029128.1 | - | 1 | 276 | 5180104 | ||
CR541908.1 | - | 1 | 141 | |||
S67156.1 | - | 1 | 299 |