Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025296.4 | NP_079572.2 | 2, 3 | 480 | ||
GenBank mRNA | AK002314.1 | - | 2, 3 | 425 | ||
AK004129.1 | - | 2, 3 | 408 | |||
AK028376.1 | - | 2, 3 | 407 | |||
BC004089.1 | - | 2, 3 | 401 | 3593736 | ||
BI734652.1 | - | 2, 3 | 480 |