Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CBS SIRNA Assays ›
          Gene Symbol:
          CBS CBSL
          Gene Name:
          cystathionine-beta-synthase
          cystathionine-beta-synthase like
          Gene Aliases:
          HIP4
          -
          Chromosome Location:
          Chr.21: 43053190 - 43076868 on Build GRCh38
          Chromosome Location:
          Chr.21: 6444868 - 6468606 on Build GRCh38
          UniGene ID:
          Hs.533013
          Hs.533013
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 7787

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CBS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000071.2 NP_000062.1 3 355
          NM_001178008.2 NP_001171479.1 3 362
          NM_001178009.2 NP_001171480.1 3 362
          NM_001320298.1 NP_001307227.1 4 356
          XM_011529777.1 XP_011528079.1 3 298
          XM_011529781.1 XP_011528083.1 4 363
          XM_011529782.1 XP_011528084.1 3 761
          XM_017028489.1 XP_016883978.1 4 758
          XM_017028490.1 XP_016883979.1 4 755
          XM_017028491.1 XP_016883980.1 3 298
          XM_017028492.1 XP_016883981.1 4 363
          GenBank mRNA AK313691.1 - 3 260
          AL531222.3 - 3 346
          BC000440.2 - 3 230 2820694
          BC010242.1 - 3 279 4154798
          BC011381.1 - 3 218 3869615
          BT007154.1 - 1 110
          DC300040.1 - 3 355
          DC315340.1 - 4 356
          HY151461.1 - 3 363
          L14577.1 - 3 268
          L19501.1 - 3 291
          X82166.1 - 3 257
          Gene Symbol: CBSL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321073.1 NP_001308002.1 3 362
          XM_011546094.1 XP_011544396.1 3 254
          XM_011546095.2 XP_011544397.1 3 396
          XM_011546097.2 XP_011544399.1 4 423
          XM_011546098.1 XP_011544400.1 3 761
          XM_011546099.1 XP_011544401.1 3 254
          XM_017028211.1 XP_016883700.1 4 759
          XM_017028212.1 XP_016883701.1 5 815
          XM_017028213.1 XP_016883702.1 3 254
          XM_017028214.1 XP_016883703.1 4 756
          XM_017028215.1 XP_016883704.1 4 371
          XM_017028216.1 XP_016883705.1 3 761
          XM_017028217.1 XP_016883706.1 3 254
          XM_017028218.1 XP_016883707.1 4 423
          GenBank mRNA BC007257.1 - 3 332 3028099

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          lyase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cysteine biosynthetic process from serine
          L-serine metabolic process
          L-serine catabolic process
          cellular amino acid biosynthetic process
          cysteine biosynthetic process via cystathionine
          cysteine biosynthetic process
          transsulfuration
          L-cysteine catabolic process
          DNA protection
          homocysteine catabolic process
          homocysteine metabolic process
          oxidation-reduction process
          hydrogen sulfide biosynthetic process
          cystathionine beta-synthase activity
          cysteine synthase activity
          protein binding
          oxygen binding
          enzyme binding
          heme binding
          pyridoxal phosphate binding
          ubiquitin protein ligase binding
          identical protein binding
          protein homodimerization activity
          metal ion binding
          nitrite reductase (NO-forming) activity
          carbon monoxide binding
          nitric oxide binding
          modified amino acid binding
          S-adenosyl-L-methionine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0